4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals
4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2500 g
Catalog number A17458.22
also known as A17458-22
Price (USD)/ Each
34.80
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
34.80
Add to cart
4-Chlorobenzophenone, 99%, Thermo Scientific Chemicals
Catalog numberA17458.22
Price (USD)/ Each
34.80
-
Add to cart
Chemical Identifiers
CAS134-85-0
IUPAC Name(4-chlorophenyl)(phenyl)methanone
Molecular FormulaC13H9ClO
InChI KeyUGVRJVHOJNYEHR-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)73.0-80.0?C
Appearance (Color)White to pale cream
Assay (GC)≥98.5%
FormPowder or lumps or flakes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only