Thermo Scientific Chemicals

Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: A17467.22
100 g, Each
Thermo Scientific Chemicals

Tetramethylammonium hexafluorophosphate, 99%, Thermo Scientific Chemicals

Catalog number: A17467.22
100 g, Each
Quantity
Catalog number: A17467.22
also known as A17467-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
558-32-7
IUPAC Name
hexafluoro-λ⁵-phosphanuide; tetramethylazanium
Molecular Formula
C4H12F6NP
InChI Key
ZFKULDQWLXAKIM-UHFFFAOYSA-N
SMILES
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
Elemental Analysis
Carbon C: 21.60-22.25% (Theory 21.93%)
Appearance (Color)
White to pale cream
Assay (unspecified)
≥98.5% by CHN analysis
Elemental Analysis
Hydrogen H: 5.44-5.60% (Theory 5.52%)
Melting Point
>300?C

Description

It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the oxidation of cytochrome c in nonaqueous solvents.

Solubility
Soluble in water.

Notes
Moisture Sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text