Thermo Scientific Chemicals

1-(2-Pyridylazo)-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: A17475.14
25 g, Each
Thermo Scientific Chemicals

1-(2-Pyridylazo)-2-naphthol, 98%, Thermo Scientific Chemicals

Catalog number: A17475.14
25 g, Each
Quantity
Catalog number: A17475.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
85-85-8
IUPAC Name
(1Z)-1-[2-(pyridin-2-yl)hydrazin-1-ylidene]-1,2-dihydronaphthalen-2-one
Molecular Formula
C15H11N3O
InChI Key
RAXUMGMWXZYADR-SDXDJHTJSA-N
SMILES
O=C1C=CC2=CC=CC=C2C1=NNC1=CC=CC=N1
Form
powder
Assay (Non-aqueous acid-base Titration)
> 98.5%
Appearance (Color)
Orange

Description

1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(2-Pyridylazo)-2-naphthol is widely used as an indicator for complexometry. It is used as a reagent for the extraction and spectrophotometric determination (for colorimetric analysis) of many transition metal elements like arsenic, chromium, cobalt, copper, cadmium, manganese, zinc and zirconium. It is also used for the analysis of uranium and thorium in solutions.

Solubility
Insoluble in water.

Notes
Incompatible with strong bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text