Thermo Scientific Chemicals

Cadmium tellurite, 99%, Thermo Scientific Chemicals

Catalog number: A17476.14
25 g, Each
Thermo Scientific Chemicals

Cadmium tellurite, 99%, Thermo Scientific Chemicals

Catalog number: A17476.14
25 g, Each
Quantity
Catalog number: A17476.14
also known as A17476-14
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15851-44-2
Form
Lumps
Appearance (Color)
Silver metallic
Assay from Supplier's CofA
≥98.5%

Description

It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the preparation, characterization and structure of Ca8[AlO2]12Te2 and Cd8[AlO2]12Te2; two new members of the sodalite family.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text