Thermo Scientific Chemicals

Creatine, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: A17477.22
100 g, Each
Thermo Scientific Chemicals

Creatine, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: A17477.22
100 g, Each
Quantity
Catalog number: A17477.22
also known as A17477-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
57-00-1
IUPAC Name
2-(N-methylcarbamimidamido)acetic acid
Molecular Formula
C4H9N3O2
InChI Key
CVSVTCORWBXHQV-UHFFFAOYSA-N
SMILES
CN(CC(O)=O)C(N)=N
Appearance (Color)
White
Identification (FTIR)
Conforms
Form
Powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Water Content (Karl Fischer Titration)
≤2%

Description

Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Creatine is involved in the rapid production of ATP associated with creatine kinases in skeletal muscle tissue. Creatine may be used as a supplement to study its mechanism of action. It acts as a nutraceutical and a neuroprotective agent.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text