Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diethylene glycol diethyl ether is used as a solvent for reactions performed at higher temperatures. It is involved in the preparation of nitrocellulose, resins and adhesives. It is utilized as a scrubbing medium to absorb carbonyl sulfide (COS), an impurity in petroleum refineries.
Solubility
Miscible with hydrocarbons, ethanol, ethyl ether and organic solvents. Immiscible with water.
Notes
Air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only
General References:
- Herreros, J. M.; Schroer, K.; Tsolakis, E. S. A. Extending the environmental benefits of ethanol-diesel blends through DGE incorporation. Appl. Energy 2015, 146, 335-343.
- Wang, W.; Yun, Z.; Tang, Z.; Gui, X. Fast Determination of Binary Vapor-Liquid Equilibrium of CO2-Based Mixtures at Elevated Pressures. J. Chem. Eng. Data 2015, 60 (6), 1642-1647.