Thermo Scientific Chemicals

Dotriacontane, 98%, Thermo Scientific Chemicals

Catalog number: A17480.06
5 g, Each
Thermo Scientific Chemicals

Dotriacontane, 98%, Thermo Scientific Chemicals

Catalog number: A17480.06
5 g, Each
Quantity
Catalog number: A17480.06
also known as A17480-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
544-85-4
IUPAC Name
dotriacontane
Molecular Formula
C32H66
InChI Key
QHMGJGNTMQDRQA-UHFFFAOYSA-N
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC
Form
Crystalline powder or flakes
Appearance (Color)
White
Assay (GC)
>97.5%

Description

Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dotriacontane is used as a solvent medium for crystallization of isotactic polypropylene.

Solubility
Soluble in hot toluene, diethyl ether, benzene and ethanol. Insoluble in water.

Notes
Incompatible with strong oxidizing agents and strong acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text