Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17484.0C
10000 g, Each
Thermo Scientific Chemicals

Sodium carbonate decahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17484.0C
10000 g, Each
Quantity
Catalog number: A17484.0C
also known as A17484-0C
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
6132-02-1
IUPAC Name
disodium decahydrate carbonate
Molecular Formula
CH20Na2O13
InChI Key
XYQRXRFVKUPBQN-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[O-]C([O-])=O
Assay (Aqueous acid-base Titration)
≥99.0 to ≤101.0%
Loss on Drying
61.75-64.09% (200?C/constant weight) (9.5-10.5 waters)
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
Colorless to white
Identification (FTIR)
Conforms

Description

Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium carbonate decahydrate is used as a raw material in various industries like glass, paper making, soap, detergent, textile and leather. It is also used as a food additive. It is also involved as a precipitation agent of carbonate as well as an analytical reagent in laboratories. It plays an important role as strong base and powerful electrolyte. It finds application in metal refining and cement production.

Solubility
Soluble in water and glycerol. Insoluble in alcohol.

Notes
Air sensitive. Incompatible with strong acids and aluminum. Keep the container tightly closed.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text