Thermo Scientific Chemicals

Ammonium magnesium phosphate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A17497.22
100 g, Each
Thermo Scientific Chemicals

Ammonium magnesium phosphate hexahydrate, 98%, Thermo Scientific Chemicals

Catalog number: A17497.22
100 g, Each
Quantity
Catalog number: A17497.22
also known as A17497-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13478-16-5
IUPAC Name
magnesium(2+) ammonium hexahydrate phosphate
Molecular Formula
H16MgNO10P
InChI Key
CKMXBZGNNVIXHC-UHFFFAOYSA-L
SMILES
[NH4+].O.O.O.O.O.O.[Mg++].[O-]P([O-])([O-])=O
Form
Powder
Identification (FTIR)
Conforms
Total Metal Impurities
Cobalt Co: ≤10ppm
Total Metal Impurities
Nickel Ni: ≤75ppm
Appearance (Color)
White

Description

It is used as pharmaceutical intermediae.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediae.

Solubility
Insoluble in water.

Notes
Store in a cool place. Ensure proper ventilation.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text