Thermo Scientific Chemicals

Sodium chromate tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17499.36
500 g, Each
Thermo Scientific Chemicals

Sodium chromate tetrahydrate, 99+%, Thermo Scientific Chemicals

Catalog number: A17499.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: A17499.36
also known as A17499-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-82-9
IUPAC Name
disodium dioxochromiumbis(olate)
Molecular Formula
CrNa2O4
InChI Key
PXLIDIMHPNPGMH-UHFFFAOYSA-N
SMILES
[Na+].[Na+].[O-][Cr]([O-])(=O)=O
Appearance (Color)
Yellow
Assay (Iodometric Titration)
≥99.0 to ≤101.0%
Form
Crystals or crystalline powder or powder

Description

Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium chromate tetrahydrate is used as a corrosion inhibitor, wood preservative and an auxiliary in textile industry. It serves as an oxidant, which is used to prepare carboxylic acids and ketones from primary and secondary alcohols respectively. Further, it is used as a starting material to prepare other chromium compounds and as a catalyst in organic synthesis especially pharmaceuticals and agro chemicals.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible withstrong reducing agents, organic materials, strong acids, combustible materials and powdered metals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text