Thermo Scientific Chemicals

Tetrahydrothiophene 1-oxide, 97%, Thermo Scientific Chemicals

Catalog number: A17502.18
50 g, Each
Thermo Scientific Chemicals

Tetrahydrothiophene 1-oxide, 97%, Thermo Scientific Chemicals

Catalog number: A17502.18
50 g, Each
Quantity
Catalog number: A17502.18
also known as A17502-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1600-44-8
IUPAC Name
1λ⁴-thiolan-1-one
Molecular Formula
C4H8OS
InChI Key
ISXOBTBCNRIIQO-UHFFFAOYSA-N
SMILES
O=S1CCCC1
Assay (GC)
≥96.0%
Refractive Index
1.5190-1.5240 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow

Description

In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.

Solubility
Fully miscible in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text