Bromocresol Green sodium salt
Bromocresol Green sodium salt
Thermo Scientific Chemicals

Bromocresol Green sodium salt

Bromocresol Green sodium salt, C21H13Br4NaO5S, CAS Number-62625-32-5, Bromocresol Green, water soluble, 100g, 263-657-4, 720.01, ∼230 deg.C (decomposition), MFCD00148898, Ambient temperatures, Soluble in water and ethanol., Yes
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number A17503.06
also known as A17503-06
Price (USD)/ Each
37.65
Online exclusive
42.10 
Save 4.45 (11%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
37.65
Online exclusive
42.10 
Save 4.45 (11%)
Add to cart
Bromocresol Green sodium salt
Catalog numberA17503.06
Price (USD)/ Each
37.65
Online exclusive
42.10 
Save 4.45 (11%)
-
Add to cart
Chemical Identifiers
CAS62625-32-5
IUPAC Namesodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular FormulaC21H13Br4NaO5S
InChI KeyHEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder
Solution Test0.1% in water: clear
Appearance (Color)Dark blue to dark green or green/brown to brown to black
Color ChangeYellow @ pH3.8 to blue @ pH5.4
lambda-max (UV-Vis)610-620nm (pH 5.4)
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Bromocresol Green sodium salt is used as a pH indicator. It is a pH sensitive triphenylmethane dye useful in various colorimetric detection studies and as an activity tracking dye for DNA agarose gel electrophoresis. It is also used in TLC (thin-layer chromatography) for visualizing compounds containing functional groups having pKa less than 5.

Solubility
Soluble in water and ethanol.

Notes
Transition interval: pH 3.8 (yellow) to pH 5.4 (blue)Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Ghaedi, M.; Khajesharifi, H.; Yadkuri, A. M.; Roosta, M.; Sahraei, R.; Daneshfar, A. Cadmium hydroxide nanowire loaded on activated carbon as efficient adsorbent for removal of Bromocresol Green. Spectrochim. Acta, Part A 2012, 86, 62-68.
  2. Chen, H.; Liao, Y. Photochromism based on reversible proton transfer. J. Photochem. Photobiol. A 2015, 300, 22-26.