Thermo Scientific Chemicals

Azure I, Thermo Scientific Chemicals

Catalog number: A17508.22
100 g, Each
Thermo Scientific Chemicals

Azure I, Thermo Scientific Chemicals

Catalog number: A17508.22
100 g, Each
Quantity
Catalog number: A17508.22
also known as A17508-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
531-55-5
IUPAC Name
3-(dimethylamino)-7-(methylamino)-5λ⁴-phenothiazin-5-ylium chloride
Molecular Formula
C15H16ClN3S
InChI Key
KFZNPGQYVZZSNV-UHFFFAOYSA-M
SMILES
[Cl-].CNC1=CC=C2N=C3C=CC(=CC3=[S+]C2=C1)N(C)C
Specific Absorbance (E 1%/1cm)
>1250 (at lambda-max)
Form
Powder
lambda-max (UV-Vis)
645 - 650nm (aqueous solution)
Appearance (Color)
Dark green or brown to black
Loss on Drying
<8% (105?C, 1 hour)

Description

Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie's modified Nocht's method for paraffin sections.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azure B is used for staining semi-thin sections of plant tissue. This dye is also useful in the NCCLS method for blood smears and Lillie′s modified Nocht′s method for paraffin sections.

Solubility
Soluble in 1 mg/ml water. Sparingly soluble in alcohol.

Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text