6-Bromopiperonal, 98%, Thermo Scientific Chemicals
6-Bromopiperonal, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

6-Bromopiperonal, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17510.06
also known as A17510-06
Price (USD)/ Each
67.90
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
67.90
Add to cart
6-Bromopiperonal, 98%, Thermo Scientific Chemicals
Catalog numberA17510.06
Price (USD)/ Each
67.90
-
Add to cart
Chemical Identifiers
CAS15930-53-7
IUPAC Name6-bromo-2H-1,3-benzodioxole-5-carbaldehyde
Molecular FormulaC8H5BrO3
InChI KeyCSQUXTSIDQURDV-UHFFFAOYSA-N
SMILESBrC1=CC2=C(OCO2)C=C1C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Free acid (titration)≤1%
Appearance (Color)White to pale cream
Assay (GC)≥97.5%
Melting Point (clear melt)126.0-132.0?C
FormPowder
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

Solubility
Soluble in DMSO, methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Christopher J. Moodya.; Graham J. Warrellow. Synthesis of the isoindolobenzazepine alkaloid lennoxamine. Tetrahedron Letters. 1987, 28, (48), 6089-6092.
  2. Frederick E. Ziegler.; Kerry W. Fowler. Substitution reactions of specifically ortho-metalated piperonal cyclohexylimine. J. Org. Chem. 1976, 41, (9), 1564-1566.