Thermo Scientific Chemicals

6-Bromopiperonal, 98%, Thermo Scientific Chemicals

Catalog number: A17510.14
25 g, Each
Thermo Scientific Chemicals

6-Bromopiperonal, 98%, Thermo Scientific Chemicals

Catalog number: A17510.14
25 g, Each
Quantity
Catalog number: A17510.14
also known as A17510-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15930-53-7
IUPAC Name
6-bromo-2H-1,3-benzodioxole-5-carbaldehyde
Molecular Formula
C8H5BrO3
InChI Key
CSQUXTSIDQURDV-UHFFFAOYSA-N
SMILES
BrC1=CC2=C(OCO2)C=C1C=O
Free acid (titration)
≤1%
Appearance (Color)
White to pale cream
Assay (GC)
≥97.5%
Melting Point (clear melt)
126.0-132.0?C
Form
Powder

Description

6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
6-Bromopiperonal is used as an intermediate in the preparation of GPR30-selective agonist G-1.

Solubility
Soluble in DMSO, methanol

Notes
Air Sensitive. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text