It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as a specialty solvent. It is a solvent for active ingredients especially DHA for sunless tanning products. It is also utilized as viscosity controlling agent.
Solubility
Soluble in water.
Notes
Store in a cool place. Ensure proper ventilation. Keep away from oxidizing agents.
RUO – Research Use Only
Hossein Zia.; Joseph K. H. Ma.; John P. O’Donnell. ; Louis A. Luzzi. Cosolvency of Dimethyl Isosorbide for Steroid Solubility.Pharm. Res. 1991, 8 (4),502-504.
Morgan Durand.; Adrien Mouret.; Valérie Molinier.; Thierry Féron.; Jean-Marie Aubry. Bitumen fluxing properties of a new class of sustainable solvents: The isosorbide di-alkyl ethers. Fuel. 2010, 89 (10),2729-2734.