Thermo Scientific Chemicals

DL-Isoleucine, 99%, Thermo Scientific Chemicals

Catalog number: A17521.22
100 g, Each
Thermo Scientific Chemicals

DL-Isoleucine, 99%, Thermo Scientific Chemicals

Catalog number: A17521.22
100 g, Each
Quantity
Catalog number: A17521.22
also known as A17521-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
443-79-8
IUPAC Name
DL-2-amino-3-methylpentanoic acid
Molecular Formula
C6H13NO2
InChI Key
AGPKZVBTJJNPAG-WHFBIAKZSA-N
SMILES
CC[C@H](C)[C@H](N)C(O)=O
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Optical Rotation
0? ±0.9? (c=1 in water)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text