Thermo Scientific Chemicals

(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine, Thermo Scientific Chemicals

Catalog number: A17523.14
25 g, Each
Thermo Scientific Chemicals

(+)-Cinchonine, 98+%, cont. up to 3% quinidine/dihydroquinidine and 3% quinine/dihydroquinine, Thermo Scientific Chemicals

Catalog number: A17523.14
25 g, Each
Quantity
Catalog number: A17523.14
also known as A17523-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-10-5
IUPAC Name
(S)-[(2R,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl](quinolin-4-yl)methanol
Molecular Formula
C19H22N2O
InChI Key
KMPWYEUPVWOPIM-FRYPIZGFNA-N
SMILES
[H][C@@]1(CN2CCC1C[C@]2([H])[C@@H](O)C1=CC=NC2=CC=CC=C12)C=C
Assay (Non-aqueous acid-base Titration)
≥98.0 to ≤102.0%
Impurity content
Quinine/Dihydroquinine ≤3.0%
Impurity content
Quinidine/Dihydroquinidine ≤3.0%
Appearance (Color)
White to pale cream
Optical Rotation
260? ? 9? (c=2, 0.2M HCl)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text