Thermo Scientific Chemicals

Diethyl phthalate, 99%, Thermo Scientific Chemicals

Catalog number: A17529.0E
2500 g, Each
Thermo Scientific Chemicals

Diethyl phthalate, 99%, Thermo Scientific Chemicals

Catalog number: A17529.0E
2500 g, Each
Quantity
Catalog number: A17529.0E
also known as A17529-0E
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
84-66-2
IUPAC Name
1,2-diethyl benzene-1,2-dicarboxylate
Molecular Formula
C12H14O4
InChI Key
FLKPEMZONWLCSK-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC=C1C(=O)OCC
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.5000-1.5035 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥98.5%

Description

Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Diethyl phthalate is used in the production of perfumes, acetyl salicylic acid, plastics and mosquito repellents. It is used in cosmetic formulations and personal care products such as sprays, nail polishes, and perfumes. It acts as an alcohol denaturant. It is also used as a camphor substitute, plasticizer in solid rocket propellants and wetting agent. It finds application as a surface lubricant in food and pharmaceutical packaging. In addition to this, it is used as a detergent base and aerosol spray.

Solubility
Miscible with ethanol, chloroform, diethyl ether, benzene, acetone, vegetable oils, ketones, carbon tetrachloride, esters, aromatic hydrocarbons and methanol. Slightly miscible with water and aliphatic solvents.

Notes
Incompatible with strong oxidizing agents, strong acids, permanganates and alkalies. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text