Thermo Scientific Chemicals

Ethyltriphenylphosphonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A17533.22
100 g, Each
Thermo Scientific Chemicals

Ethyltriphenylphosphonium iodide, 98+%, Thermo Scientific Chemicals

Catalog number: A17533.22
100 g, Each
Quantity
Catalog number: A17533.22
also known as A17533-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4736-60-1
IUPAC Name
ethyltriphenylphosphanium iodide
Molecular Formula
C20H20IP
InChI Key
SLAFUPJSGFVWPP-UHFFFAOYSA-M
SMILES
[I-].CC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Assay (Titration ex Iodide)
≥98.0 to ≤102.0%
Water Content (Karl Fischer Titration)
≤1%
Form
Crystals or powder or crystalline powder or fused solid
Appearance (Color)
White to cream
Identification (FTIR)
Conforms

Description

Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyltriphenylphosphonium iodide is involved in synthesis of diarylmethine derivatives, phosphonium salts, and bismuth(III) polynuclear halide complexes, asymmetric hydrogenation.

Solubility
Soluble in water.

Notes
Light Sensitive & Hygroscopic. Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text