Thermo Scientific Chemicals

Polyethylene glycol 6,000, Thermo Scientific Chemicals

Catalog number: A17541.30
250 g, Each
Thermo Scientific Chemicals

Polyethylene glycol 6,000, Thermo Scientific Chemicals

Catalog number: A17541.30
250 g, Each
Quantity
Catalog number: A17541.30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
25322-68-3
Molecular Formula
(C2H4O)n
InChI Key
LYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES
[H]OCCO
Hydroxyl Value
17-21mg KOH/g
Viscosity
200-270 mPas (50%, Water @ 20?C)
Appearance (Color)
White to cream
Comment
Average mol weight 5400 - 6600 (based on OH value)
Melting Point (clear melt)
55-63?C

Description

Polyethylene glycol is used in the modification of therapeutic proteins and peptides to enhance its solubility. It is also used in the fabrication of bioactive and immuno isolating barriers for encapsulation of cells. In Gas chromatorgraphy, it is used as a polar stationary phase. It is used as mold release agent and lubricant in fabricating elastomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text