Thermo Scientific Chemicals

Isopropyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A17551.22
100 g, Each
Thermo Scientific Chemicals

Isopropyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Catalog number: A17551.22
100 g, Each
Quantity
Catalog number: A17551.22
also known as A17551-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18144-43-9
IUPAC Name
propan-2-yl 4-aminobenzoate
Molecular Formula
C10H13NO2
InChI Key
JWCPZKNBPMSYND-UHFFFAOYSA-N
SMILES
CC(C)OC(=O)C1=CC=C(N)C=C1
Appearance (Color)
Pale cream to cream to pale brown
Assay (GC)
≥97.5%
Melting Point (clear melt)
80.0-87.0?C
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text