Thermo Scientific Chemicals

1,4-Benzodioxane, 98%, Thermo Scientific Chemicals

Catalog number: A17558.09
10 g, Each
Thermo Scientific Chemicals

1,4-Benzodioxane, 98%, Thermo Scientific Chemicals

Catalog number: A17558.09
10 g, Each
Quantity
Catalog number: A17558.09
also known as A17558-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
493-09-4
IUPAC Name
2,3-dihydro-1,4-benzodioxine
Molecular Formula
C8H8O2
InChI Key
BNBQRQQYDMDJAH-UHFFFAOYSA-N
SMILES
C1COC2=CC=CC=C2O1
Form
Liquid
Refractive Index
1.5475-1.5515 @ 20?C
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%

Description

Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo-1,4-dioxane (2,3-dihydro-1,4-benzodioxin) is used in the synthesis of stereoisomers which were evaluated as α- and β- adrenergic antagonists

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text