Thermo Scientific Chemicals

N-Acetyl-DL-tryptophan, 99%, Thermo Scientific Chemicals

Catalog number: A17562.22
100 g, Each
Thermo Scientific Chemicals

N-Acetyl-DL-tryptophan, 99%, Thermo Scientific Chemicals

Catalog number: A17562.22
100 g, Each
Quantity
Catalog number: A17562.22
also known as A17562-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
87-32-1
IUPAC Name
2-acetamido-3-(1H-indol-3-yl)propanoic acid
Molecular Formula
C13H14N2O3
InChI Key
DZTHIGRZJZPRDV-UHFFFAOYSA-N
SMILES
CC(=O)NC(CC1=CNC2=CC=CC=C12)C(O)=O
Assay (Aqueous acid-base Titration)
≥98.5 to ≤101.5%
Appearance (Color)
White to pale cream
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥98.5%

Description

N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Acetyl-DL-tryptophan is produced via chemical synthesis using the standard amino acid, tryptophan. It can be used in downstream applications of biopharmaceutical production as a transfer agent or stabilizer.

Solubility
Soluble in 1N Sodium Hydroxide, water, and methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text