Thermo Scientific Chemicals

Alginic acid, Thermo Scientific Chemicals

Catalog number: A17582.36
500 g, Each
Thermo Scientific Chemicals

Alginic acid, Thermo Scientific Chemicals

Catalog number: A17582.36
500 g, Each
Quantity
Catalog number: A17582.36
also known as A17582-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
9005-32-7
Molecular Formula
(C6H8O7)A(C6H8O7)B
SMILES
O[C@H]1[C@@H](O)[C@H](O-*)[C@H](O[C@H]1O-*)C(O)=O.O[C@H]1[C@@H](O)[C@H](O-*)[C@@H](O[C@H]1O-*)C(O)=O
Molecular Weight (g/mol)
NaN
Loss on Drying
≤15%
Appearance (Color)
White to cream
Assay (Aqueous acid-base Titration)
19-25% w/w COOH (dried material)
Form
powder

Description

A straight chain, hydrophilic, colloidal polyuronic acidAlginic acid is used as a precursor for the preparation of sodium alginate, potassium alginate and calcium alginate, which finds application in various industries such as food, textile printing and pharmaceuticals. It acts as an additive in dehydrated products such as slimming aids. It is also used in the production of paper and textiles. It is actively involved in waterproofing, fireproofing fabrics. It is utilized as a thickening agent in drinks, ice cream and cosmetics. It plays an important role as a raw material for the synthesis of propylene glycol alginate and polysaccharide sulfate. It is also useful to treat and purify waste water having strong absorbility.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text