Thermo Scientific Chemicals

Triisopropyl borate, 98+%, Thermo Scientific Chemicals

Catalog number: A17592.AE
100 mL, Each
Thermo Scientific Chemicals

Triisopropyl borate, 98+%, Thermo Scientific Chemicals

Catalog number: A17592.AE
100 mL, Each
Quantity
Catalog number: A17592.AE
also known as A17592-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5419-55-6
IUPAC Name
tris(propan-2-yl) borate
Molecular Formula
C9H21BO3
InChI Key
NHDIQVFFNDKAQU-UHFFFAOYSA-N
SMILES
CC(C)OB(OC(C)C)OC(C)C
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥98.0%
Refractive Index
1.3740-1.3780 @ 20?C
Identification (FTIR)
Conforms

Description

Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triisopropyl borate is used as reagent in Pd-catalyzed coupling reaction with aryl halides such as Suzuki reaction. It is used as a reagent for the preparation of the boronic acids and esters; as a Lewis acid catalyst and involved in the ortho-borylation of 1-substituted naphthalenes. Furthermore, it plays an important role as a catalyst for the production of resins, waxes, paints and varnishes.

Solubility
Miscible with ether, ethanol, isopropanol and benzene.

Notes
Moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Decomposes in water.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text