Thermo Scientific Chemicals

Methylcyclopentadiene dimer, tech. 90%, stab. with 200ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A17596.AP
500 mL, Each
Thermo Scientific Chemicals

Methylcyclopentadiene dimer, tech. 90%, stab. with 200ppm 4-tert-butylcatechol, Thermo Scientific Chemicals

Catalog number: A17596.AP
500 mL, Each
Quantity
Catalog number: A17596.AP
also known as A17596-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
26472-00-4
IUPAC Name
4,9-dimethyltricyclo[5.2.1.0²,⁶]deca-3,8-diene
Molecular Formula
C12H16
InChI Key
IYQYZZHQSZMZIG-UHFFFAOYNA-N
SMILES
CC1=CC2C(C1)C1CC2C(C)=C1
Appearance (Color)
Clear colorless to yellow
Assay (GC)
≥88.0% (total isomers)
Refractive Index
1.4940-1.5020 @ 20?C
Comment
Stabilised with 200ppm 4-tert-butylcatechol
Form
Liquid

Description

Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylcyclopentadiene dimer is used to study the competition with ethylene for binding and biological activities. Used in the synthesis of rubbers and resins intended for use in contact with food or drink.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text