Thallium(III) oxide, 96%, Thermo Scientific Chemicals
Thallium(III) oxide, 96%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Thallium(III) oxide, 96%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number A17601.09
also known as A17601-09
Price (USD)/ Each
63.70
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
63.70
Add to cart
Thallium(III) oxide, 96%, Thermo Scientific Chemicals
Catalog numberA17601.09
Price (USD)/ Each
63.70
-
Add to cart
Chemical Identifiers
CAS1314-32-5
IUPAC Nameoxo[(oxothallanyl)oxy]thallane
Molecular FormulaO3Tl2
InChI KeyQTQRFJQXXUPYDI-UHFFFAOYSA-N
SMILESO=[Tl]O[Tl]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Brown
FormCrystals or powder or crystalline powder
Assay from Suppliers CofA≥95% (U.S. specification)
Assay (special)≥94.0 to ≤106.0% by Thallium EA (non-U.S. specification)
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thallium(III) oxide is used in organic synthesis and as an analytical reagent

Solubility
Insoluble in water and alkalis. Decomposes in HCl and H2SO4

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Robert A. Van Leeuwen.; Chen-Jen Hung.; Daniel R. Kammler.; Jay A. Switzer. Optical and Electronic Transport Properties of Electrodeposited Thallium(III) Oxide Films. J. Phys. Chem. 1995, 99, (41), 15247-15252.
  2. A. D. Berry.; R. T. Holm.; R. L. Mowery.; N. H. Turner.; M. Fatemi. Growth and characterization of thin films of thallium(III) oxide by organometallic chemical vapor deposition. Chem. Mater. 1991, 3, (1), 72-77.