Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals
Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2500 g
100 g
500 g
Catalog number A17615.0E
also known as A17615-0E
Price (USD)/ Each
278.65
Online exclusive
309.00 
Save 30.35 (10%)
-
Add to cart
Quantity:
2500 g
Request bulk or custom format
Price (USD)/ Each
278.65
Online exclusive
309.00 
Save 30.35 (10%)
Add to cart
Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals
Catalog numberA17615.0E
Price (USD)/ Each
278.65
Online exclusive
309.00 
Save 30.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10034-96-5
IUPAC Namemanganese(2+) hydrate sulfate
Molecular FormulaH2MnO5S
InChI KeyISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILESO.[Mn++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale pink
Complexometric Titration≥98.5 to ≤101.5%
CommentMaterial Sourced in UK and US
Loss on Drying5.63-15.18% (450?C/2hrs) (0.5-1.5 waters) (UK Sourced Material)
FormCrystals or powder or crystalline powder
View more
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Wang, D.; Belharouak, I.; Ortega, L. H.; Zhang, X.; Xu, R.; Zhou, D.; Zhou, G.; Amine, K. Synthesis of high capacity cathodes for lithium-ion batteries by morphology-tailored hydroxide co-precipitation. J. Power Sources 2015, 274, 451-457.
  2. Raza, W.; Khan, A.; Alam, U.; Muneer, M.; Bahnemann, D. Facile fabrication of visible light induced Bi2O3 nanorod using conventional heat treatment method. J. Mol. Struct. 2015, 1107, 39-46.