Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A17615.36
500 g, Each
Thermo Scientific Chemicals

Manganese(II) sulfate monohydrate, 99%, Thermo Scientific Chemicals

Catalog number: A17615.36
500 g, Each
Quantity
Catalog number: A17615.36
also known as A17615-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-96-5
IUPAC Name
manganese(2+) hydrate sulfate
Molecular Formula
H2MnO5S
InChI Key
ISPYRSDWRDQNSW-UHFFFAOYSA-L
SMILES
O.[Mn++].[O-]S([O-])(=O)=O
Appearance (Color)
White to pale pink
Complexometric Titration
≥98.5 to ≤101.5%
Comment
Material Sourced in UK and US
Loss on Drying
5.63-15.18% (450?C/2hrs) (0.5-1.5 waters) (UK Sourced Material)
Form
Crystals or powder or crystalline powder

Description

Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Manganese(II) sulfate monohydrate acts as a colorant in dyes. It is used in the preparation of manganese and manganese dioxide. It finds application for remediation of manganese-deficient soil. It is also used in dry-cell batteries.

Solubility
Soluble in water and alcohol. Insoluble in ether.

Notes
Loses all water at 400-450°. Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text