Thermo Scientific Chemicals

Adenine hydrochloride, 98+%, cont. up to ca 5% water

Catalog number: A17622.22
100 g, Each
Thermo Scientific Chemicals

Adenine hydrochloride, 98+%, cont. up to ca 5% water

Catalog number: A17622.22
100 g, Each
Quantity
Catalog number: A17622.22
also known as A17622-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2922-28-3
IUPAC Name
7H-purin-6-amine hydrochloride
Molecular Formula
C5H6ClN5
InChI Key
UQVDQSWZQXDUJB-UHFFFAOYSA-N
SMILES
Cl.NC1=C2NC=NC2=NC=N1
Assay (Titration ex Chloride)
>98.0 to <102.0% (Dry wt. basis)
Appearance (Color)
White
Water Content (Karl Fischer Titration)
< 7.5% (hemihydrate = 5.0%)
Form
Powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text