Thermo Scientific Chemicals

2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals

Catalog number: A17625.18
50 g, Each
Thermo Scientific Chemicals

2-Bromoethylamine hydrobromide, 98+%, Thermo Scientific Chemicals

Catalog number: A17625.18
50 g, Each
Quantity
Catalog number: A17625.18
also known as A17625-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2576-47-8
IUPAC Name
hydrogen 2-bromoethan-1-amine bromide
Molecular Formula
C2H7Br2N
InChI Key
WJAXXWSZNSFVNG-UHFFFAOYSA-N
SMILES
[H+].[Br-].NCCBr
Appearance (Color)
White to pale brown
Assay (Titration ex Bromide)
≥98.0 to ≤102.0%
Identification (FTIR)
Conforms
Form
Crystals or crystalline powder
Assay (Freebase GC)
≥98.0%

Description

2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Bromoethylamine hydrobromide is used to construct C2-symmetric imidazolidinylidene ligands with a dioxolane backbone. It acts as a building block which is used for proteomics research. It is also used as organic and pharmaceutical intermediates for the preparation of first-aid medicines.

Solubility
Soluble in water.

Notes
Hygroscopic and moisture sensitive. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text