Thermo Scientific Chemicals

Benzyl bromoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A17630.18
50 g, Each
Thermo Scientific Chemicals

Benzyl bromoacetate, 97%, Thermo Scientific Chemicals

Catalog number: A17630.18
50 g, Each
Quantity
Catalog number: A17630.18
also known as A17630-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
5437-45-6
IUPAC Name
benzyl 2-bromoacetate
Molecular Formula
C9H9BrO2
InChI Key
JHVLLYQQQYIWKX-UHFFFAOYSA-N
SMILES
BrCC(=O)OCC1=CC=CC=C1
Refractive Index
1.5415-1.5465 @ 20?C
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥96.0%
Identification (FTIR)
Conforms
Form
Liquid

Description

It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in biological research purpose. Benzyl bromoacetate was used in the alkylation of (-)-2,3-O-isopropylidene-D-threitol that afforded lipopeptide, 2-[(4R,5R)-5-({[(9H-fluoren-9-yl)methoxy]carbonylaminomethyl}-2,2-dimethyl-1,3-dioxolan-4-yl)methoxy]acetic acid.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place. Store under 20˚C Freezer, Under Inert Atmosphere.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text