Thermo Scientific Chemicals

Hexamethylditin, 97%, Thermo Scientific Chemicals

Catalog number: A17638.06
5 g, Each
Thermo Scientific Chemicals

Hexamethylditin, 97%, Thermo Scientific Chemicals

Catalog number: A17638.06
5 g, Each
Quantity
Catalog number: A17638.06
also known as A17638-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
661-69-8
IUPAC Name
hexamethyldistannane
Molecular Formula
C6H18Sn2
InChI Key
CCRMAATUKBYMPA-UHFFFAOYSA-N
SMILES
C[Sn](C)(C)[Sn](C)(C)C
Assay (GC)
≥96.0%
Refractive Index
1.5385-1.5435 @ 20°C
Identification (FTIR)
Conforms
Appearance (Color)
Colorless to pale yellow
Form
Clear to hazy or turbid liquid

Description

It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to prepare aryl tin compounds for microwave-assisted Stille cross-coupling with halo pyridines or copper-mediated O-arylation of phenols.

Solubility
Not miscible or difficult to mix in water.

Notes
Air Sensitive. Keep away from oxidizing agents, heat and air. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text