Thermo Scientific Chemicals

2-Methoxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A17650.0B
1000 g, Each
Thermo Scientific Chemicals

2-Methoxynaphthalene, 98%, Thermo Scientific Chemicals

Catalog number: A17650.0B
1000 g, Each
Quantity
Catalog number: A17650.0B
also known as A17650-0B
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-04-9
IUPAC Name
2-methoxynaphthalene
Molecular Formula
C11H10O
InChI Key
LUZDYPLAQQGJEA-UHFFFAOYSA-N
SMILES
COC1=CC=C2C=CC=CC2=C1
Form
Crystals or powder or crystalline powder
Appearance (Color)
White to yellow-brown
Melting Point (clear melt)
70.0-77.0°C
Assay (GC)
≥97.5%

Description

2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methoxynaphthalene acylation is used as a model reaction to study the catalytic benefits of delamination. It was also used to study the alkali-metal-mediated manganation (AMMMn) reactions.

Solubility
Soluble in benzene, carbon disulfide, diethyl ether. Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text