Dimethyltin dichloride, 98%, Thermo Scientific Chemicals
Dimethyltin dichloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyltin dichloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number A17661.06
also known as A17661-06
Price (USD)/ Each
75.20
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
75.20
Add to cart
Dimethyltin dichloride, 98%, Thermo Scientific Chemicals
Catalog numberA17661.06
Price (USD)/ Each
75.20
-
Add to cart
Chemical Identifiers
CAS753-73-1
Molecular FormulaC2H8Cl2Sn
InChI KeyQLQTYMCOTSXVHH-UHFFFAOYSA-N
SMILESCl.Cl.C[SnH2]C
Molecular Weight (g/mol)221.70
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White
Melting Point (clear melt)104.0-110.0?C
Assay (Titration ex Chloride)≥97.5 to ≤102.5%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only