Chemical Identifiers
CAS37558-29-5
IUPAC Name2,5-dioxopyrrolidin-1-yl 1-hydroxy-2,2,5,5-tetramethyl-2,5-dihydro-1H-pyrrole-3-carboxylate
Molecular FormulaC13H18N2O5
InChI KeyVDSVTMQVXNIMMJ-UHFFFAOYSA-N
SMILESCC1(C)C=C(C(=O)ON2C(=O)CCC2=O)C(C)(C)N1O
View more