Thermo Scientific Chemicals

3-Nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A17670.22
100 g, Each
Thermo Scientific Chemicals

3-Nitrobenzenesulfonyl chloride, 98%, Thermo Scientific Chemicals

Catalog number: A17670.22
100 g, Each
Quantity
Catalog number: A17670.22
also known as A17670-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
121-51-7
IUPAC Name
3-nitrobenzene-1-sulfonyl chloride
Molecular Formula
C6H4ClNO4S
InChI Key
MWWNNNAOGWPTQY-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=CC(=C1)S(Cl)(=O)=O
Appearance (Color)
Pale cream to cream
Form
Crystals or powder or crystalline powder
Assay (GC)
≥97.5%
Assay (Titration ex Chloride)
≥97.5 to ≤102.5%
Melting Point (clear melt)
58.0-65.0?C

Description

It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a biochemical for proteomics research. It is also used in the preparation of: acyl-2-aminobenzimidazole analogs and 6-chloro-2-methyl-3-{1-[(3-nitrophenyl)sulfonyl]-1H-pyrazol-3-yl}imidazo[1,2-a]pyridine.

Solubility
Hydrolyzes slowly in water.

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text