3,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
3,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
25 g
100 g
250 g
Catalog number A17672.18
also known as A17672-18
Price (USD)/ Each
46.30
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
46.30
Add to cart
3,4-Dichlorobenzyl chloride, 98%, Thermo Scientific Chemicals
Catalog numberA17672.18
Price (USD)/ Each
46.30
-
Add to cart
Chemical Identifiers
CAS102-47-6
IUPAC Name1,2-dichloro-4-(chloromethyl)benzene
Molecular FormulaC7H5Cl3
InChI KeyYZIFVWOCPGPNHB-UHFFFAOYSA-N
SMILESClCC1=CC=C(Cl)C(Cl)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
Refractive Index1.5755-1.5795 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only