Stearic acid, 90+%, Thermo Scientific Chemicals
Stearic acid, 90+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Stearic acid, 90+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5000 g
1000 g
Catalog number A17673.0I
also known as A17673-0I
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Quantity:
5000 g
Request bulk or custom format
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
Add to cart
Stearic acid, 90+%, Thermo Scientific Chemicals
Catalog numberA17673.0I
Price (USD)/ Each
94.65
Online exclusive
105.00 
Save 10.35 (10%)
-
Add to cart
Chemical Identifiers
CAS57-11-4
IUPAC Nameoctadecanoic acid
Molecular FormulaC18H36O2
InChI KeyQIQXTHQIDYTFRH-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (Silylated GC)≥90.0%
FormFlakes
Melting Point66-71?C (≤2?C range)
Appearance (Color)White
Identification (FTIR)Conforms
Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Stearic acid is used as a raw material in the production of detergents, surfactants, soaps, and cosmetics such as shampoos and shaving cream products. Its salts are used to soften polyvinyl chloride and a component of grease. It is used for the preparation of softeners in textile sizing with castor oil and dietary supplements. It acts as a hardener in candles with simple sugar or corn syrup. It is utilized for the coating of metal powders like aluminum and iron in fireworks. It finds application as a common lubricant during injection molding and pressing of ceramic powders. It acts as a key raw material for the synthesis of stearyl alcohol by reduction.

Solubility
Soluble in ethanol, dimethyl sulfoxide, dimethyl formamide, carbon terachloride, toluene, chloroform, acetone and carbondisulfide. Insoluble in water.

Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
RUO – Research Use Only

General References:

  1. Cruz, L.; Fernandes, V. C.; Araújo, P.; Mateus, N.; de Freitas, V. Synthesis, characterisation and antioxidant features of procyanidin B4 and malvidin-3-glucoside stearic acid derivatives. Food Chem. 2015, 174, 480-486.
  2. Sarı, A.; Alkan, C.; Özcan, A. N. Synthesis and characterization of microano capsules of PMMA/capric-stearic acid eutectic mixture for low temperature-thermal energy storage in buildings. Energy Build. 2015, 90, 106-113.