Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17676.14
25 g, Each
Thermo Scientific Chemicals

2,3,4,5,6-Pentafluorobenzenesulfonyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17676.14
25 g, Each
Quantity
Catalog number: A17676.14
also known as A17676-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
832-53-1
IUPAC Name
pentafluorobenzene-1-sulfonyl chloride
Molecular Formula
C6ClF5O2S
InChI Key
UOJCTEGNHXRPKO-UHFFFAOYSA-N
SMILES
FC1=C(F)C(F)=C(C(F)=C1F)S(Cl)(=O)=O
Appearance (Color)
Clear colorless to pale yellow
Assay (GC)
≥98.0%
Refractive Index
1.4765-1.4805 @ 20?C
Form
Liquid

Description

Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pentafluorobenzenesulfonyl chloride is used as derivatizing reagent for simultaneous determination of fluoxetine and p-trifluoromethylphenol, an O-dealkylated metabolite of fluoxetine in human liver microsomes. It is also used for electrophore labeling of small tyrosyl peptides for their analysis by gas chromatography with electron-capture detection. It is also employed as derivatizing reagent for amphetamine in determination of amphetamine in human plasma samples using gas chromatography with electron-capture detection.

Solubility
Soluble in water (reacts slowly).

Notes
Moisture Sensitive. Store away from oxidizing agents, active metals and bases, water/moisture. Keep the container tightly closed in a cool, dry place. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text