Thermo Scientific Chemicals

Guaiazulene, 98+%, Thermo Scientific Chemicals

Catalog number: A17690.06
5 g, Each
Thermo Scientific Chemicals

Guaiazulene, 98+%, Thermo Scientific Chemicals

Catalog number: A17690.06
5 g, Each
Quantity
Catalog number: A17690.06
also known as A17690-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
489-84-9
IUPAC Name
1,4-dimethyl-7-(propan-2-yl)azulene
Molecular Formula
C15H18
InChI Key
FWKQNCXZGNBPFD-UHFFFAOYSA-N
SMILES
CC(C)C1=CC=C(C)C2=CC=C(C)C2=C1
Appearance (Color)
Blue to dark blue
Assay from Supplier's CofA
≥98.0% (US-sourced material)
Form
Crystals or fused solid
Comment
Material sourced in UK and US
Assay (GC)
≥98.0% (UK-sourced material)

Description

It is employed as antimicrobial agent and perfuming agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as antimicrobial agent and perfuming agent in cosmetics industry.

Solubility
Soluble in alcohol, water, 0.1115 mg/L @ 25°C (est).

Notes
Store in dark. Protect from heat, oxidizing agents and heat. Keep it refrigerated.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text