8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only
Scott A. Hilderbrand.; Ralph Weissleder. One-pot synthesis of new symmetric and asymmetric xanthene dyes.Tetrahedron Lett.2007,48(25),4383-4385.
Jin Young Noh.; Soojin Kim.; In Hong Hwang.; Ga Ye Lee.; Juhye Kang.; So Hyun Kim.; Jisook Min.; Sungsu Park.; Cheal Kim.; Jinheung Kim. Solvent-dependent selective fluorescence assay of aluminum and gallium ions using julolidine-based probe.Dyes and Pigments,.2013,99(3), 1016-1021.