Thermo Scientific Chemicals

8-Hydroxyjulolidine, 97%, Thermo Scientific Chemicals

Catalog number: A17692.06
5 g, Each
Thermo Scientific Chemicals

8-Hydroxyjulolidine, 97%, Thermo Scientific Chemicals

Catalog number: A17692.06
5 g, Each
Quantity
Catalog number: A17692.06
also known as A17692-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
41175-50-2
IUPAC Name
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol
Molecular Formula
C12H15NO
InChI Key
FOFUWJNBAQJABO-UHFFFAOYSA-N
SMILES
OC1=CC=C2CCCN3CCCC1=C23
Appearance (Color)
Pale brown to brown or pink
Assay (GC)
≥96.0%
Melting Point (clear melt)
129.0-139.0?C
Form
Powder

Description

8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.

Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text