Ammonium sulfamate, 98+%, Thermo Scientific Chemicals
Ammonium sulfamate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium sulfamate, 98+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number A17696.30
also known as A17696-30
Price (USD)/ Each
42.65
Online exclusive
47.00 
Save 4.35 (9%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
47.00 
Save 4.35 (9%)
Add to cart
Ammonium sulfamate, 98+%, Thermo Scientific Chemicals
Catalog numberA17696.30
Price (USD)/ Each
42.65
Online exclusive
47.00 
Save 4.35 (9%)
-
Add to cart
Chemical Identifiers
CAS7773-06-0
IUPAC Nameammonium sulfamate
Molecular FormulaH6N2O3S
InChI KeyGEHMBYLTCISYNY-UHFFFAOYSA-N
SMILES[NH4+].NS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
FormCrystals or powder or crystalline powder or fused solid
Water Content (Karl Fischer Titration)≤1%
Appearance (Color)White
Assay (Non-aqueous acid-base Titration)≥98.0 to ≤102.0%
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ammonium sulfamate is applied in water solution or oil-water emulsion as a leaf, or foliar spray for control of woody plants, or it is applied as crystals or concentrated solution to cuts in the bark or on freshly-cut stumps of undesirable trees to prevent resprouting. Used as a compost accelerator, flame retardant and in industrial processes.

Solubility
Soluble in water(2800g/L ). Insoluble in methanol, ether, n-octanol.

Notes
Hygroscopic. Store at room temperature. Stored in airtight containers which are kept closed and away from air and moisture.
RUO – Research Use Only