Thermo Scientific Chemicals

2,4-Diaminophenol dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17697.14
25 g, Each
Thermo Scientific Chemicals

2,4-Diaminophenol dihydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: A17697.14
25 g, Each
Quantity
Catalog number: A17697.14
also known as A17697-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
137-09-7
IUPAC Name
dihydrogen 2,4-diaminophenol dichloride
Molecular Formula
C6H10Cl2N2O
InChI Key
KQEIJFWAXDQUPR-UHFFFAOYSA-N
SMILES
[H+].[H+].[Cl-].[Cl-].NC1=CC=C(O)C(N)=C1
Assay (Titration ex Chloride)
≥98.0 to ≤102.0%
Appearance (Color)
Brown to pale brown to grey
Form
Crystals or powder or crystalline powder

Description

2,4-Diaminophenol dihydrochloride (Amidol) is used in the dye manufacturing industries and as a color accelerator in photographic developers. It is also employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4-Diaminophenol dihydrochloride (Amidol) is used in the dye manufacturing industries and as a color accelerator in photographic developers. It is also employed as a pharmaceutical intermediate.

Solubility
Soluble in water; very faint turbidity [27.5 g/100 mL (15°C).]

Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text