Thermo Scientific Chemicals

1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A17705.36
500 g, Each
Thermo Scientific Chemicals

1-Bromo-4-nitrobenzene, 98%, Thermo Scientific Chemicals

Catalog number: A17705.36
500 g, Each
Quantity
Catalog number: A17705.36
also known as A17705-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
586-78-7
IUPAC Name
1-bromo-4-nitrobenzene
Molecular Formula
C6H4BrNO2
InChI Key
ZDFBKZUDCQQKAC-UHFFFAOYSA-N
SMILES
[O-][N+](=O)C1=CC=C(Br)C=C1
Assay (GC)
≥97.5%
Appearance (Color)
White to cream to orange to brown
Form
Powder
Identification (FTIR)
Conforms
Melting Point (clear melt)
122.0-128.0?C

Description

It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be employed as a syntheses material intermediate. It is also used as a pharmaceutical intermediate.

Solubility
Insoluble in water. Almost transparent in hot ethyl alcohol. Soluble in ether

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed in a cool, dry place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text