Thermo Scientific Chemicals

D-Tyrosine, 99%, Thermo Scientific Chemicals

Catalog number: A17709.06
5 g, Each
Thermo Scientific Chemicals

D-Tyrosine, 99%, Thermo Scientific Chemicals

Catalog number: A17709.06
5 g, Each
Quantity
Catalog number: A17709.06
also known as A17709-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
556-02-5
IUPAC Name
(2R)-2-amino-3-(4-hydroxyphenyl)propanoic acid
Molecular Formula
C9H11NO3
InChI Key
OUYCCCASQSFEME-SVGMAFHSNA-N
SMILES
N[C@H](CC1=CC=C(O)C=C1)C(O)=O
Form
Powder
Optical Rotation
11.0 ± 1.0? (C=4 in 1N HCl)
Appearance (Color)
White
Assay (Non-aqueous acid-base Titration)
≥98.5 to ≤101.5%

Description

D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Tyrosine is synthesized from phenylalanine. It is also the precursor of epinephrine, thyroid hormones, and melanin.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text