Thermo Scientific Chemicals

Cyclopentanecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A17710.03
1 g, Each
Thermo Scientific Chemicals

Cyclopentanecarbonitrile, 98%, Thermo Scientific Chemicals

Catalog number: A17710.03
1 g, Each
Quantity
Catalog number: A17710.03
also known as A17710-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4254-02-8
IUPAC Name
cyclopentanecarbonitrile
Molecular Formula
C6H9N
InChI Key
SVPZJHKVRMRREG-UHFFFAOYSA-N
SMILES
N#CC1CCCC1
Assay (GC)
≥97.5%
Form
Liquid
Refractive Index
1.4410-1.4450 @ 20?C
Appearance (Color)
Clear colorless

Description

Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentanecarbonitrile is used as a substrate and reagent to investigate the specific activity of nitrile and amide hydrolyzing enzymes isolated from candida guilliermondii cells. It is also used to prepare cyclopentanecarboxamide by using tetrabutylammonium hydrogen sulfate as a catalyst in the presence of reagents such as aqueous hydrogen peroxide and sodium hydroxide.

Solubility
Miscible with toluene.

Notes
Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text