Thermo Scientific Chemicals

2,3-Dibromopropionyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A17712.09
10 g, Each
Thermo Scientific Chemicals

2,3-Dibromopropionyl chloride, 95%, Thermo Scientific Chemicals

Catalog number: A17712.09
10 g, Each
Quantity
Catalog number: A17712.09
also known as A17712-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
18791-02-1
IUPAC Name
2,3-dibromopropanoyl chloride
Molecular Formula
C3H3Br2ClO
InChI Key
HWKWYDXHMQQDQJ-UHFFFAOYNA-N
SMILES
ClC(=O)C(Br)CBr
Assay (Methyl Ester GC)
>94.0%
Form
Liquid
Appearance (Color)
Clear colorless to yellow

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text